Learn More
Medchemexpress LLC Valproic acid-d15 | 362049-65-8 | 99.8% | 159.30 g·mol⁻¹ | C8HD15O2 | 10 MG
SDP
Catalog No. 5000243141
Encompass_Preferred
Valproic acid-d15 is the deuterium-labeled stable isotope of valproic acid provided as a high-purity analytical standard for mass spectrometry, isotope-dilution quantification, and pharmacokinetic or metabolic tracer studies. It offers consistent isotopic labeling for accurate internal standard performance.
  • Deuterium-labeled (d15) analog for isotope-dilution assays.
  • Purity 99.8% for reliable analytical results.
  • Suitable as an internal standard for LC-MS/MS workflows.
  • Useful for tracer studies and pharmacokinetic/metabolic research.
  • Available in small, pre-weighed quantities for method development.
Catalog No. 50-002-43141 Supplier Medchemexpress LLC Supplier No. HY10585S210MG
Valproic acid-d15 is the deuterium-labeled stable isotope of valproic acid provided as a high-purity analytical standard for mass spectrometry, isotope-dilution quantification, and pharmacokinetic or metabolic tracer studies. It offers consistent isotopic labeling for accurate internal standard performance.
  • Deuterium-labeled (d15) analog for isotope-dilution assays.
  • Purity 99.8% for reliable analytical results.
  • Suitable as an internal standard for LC-MS/MS workflows.
  • Useful for tracer studies and pharmacokinetic/metabolic research.
  • Available in small, pre-weighed quantities for method development.