Learn More
Medchemexpress LLC Valproic acid-d4 10mg | 87745-17-3 | 10MG
SDP
Catalog No. 5000244416
Encompass_Preferred
Valproic acid-d4 is the deuterium labeled Valproic acid Valproic acid (VPA 2-Propylpentanoic Acid) is an HDAC inhibitor with IC50 in the range of 0 5 and 2 mM also inhibits HDAC1 (IC50 400 M) and induces proteasomal degradation of HDAC2 Valproic acid activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells Valproic acid sodium salt is used in the treatment of epilepsy bipolar disorder and prevention of migraine headaches[1 [2
Catalog No. 50-002-44416 Supplier Medchemexpress LLC Supplier No. HY10585S10MG
Valproic acid-d4 is the deuterium labeled Valproic acid Valproic acid (VPA 2-Propylpentanoic Acid) is an HDAC inhibitor with IC50 in the range of 0 5 and 2 mM also inhibits HDAC1 (IC50 400 M) and induces proteasomal degradation of HDAC2 Valproic acid activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells Valproic acid sodium salt is used in the treatment of epilepsy bipolar disorder and prevention of migraine headaches[1 [2