Learn More
Cayman Chemical ValproIc AcId-d4 10mg
SDP
Catalog No. 502945819
Encompass_Preferred
An internal standard for the quantification of valproic acid by GC- or LC-MS
Catalog No. 50-294-5819 Supplier Cayman Chemical Supplier No. 3996610 MG
An internal standard for the quantification of valproic acid by GC- or LC-MS