Learn More
Cayman Chemical Valproic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 87745-17-3, 5 mg
SDP
Catalog No. 502942844
Encompass_Preferred
An internal standard for the quantification of valproic acid by GC- or LC-MS
Catalog No. 50-294-2844 Supplier Cayman Chemical Supplier No. 399665 MG
An internal standard for the quantification of valproic acid by GC- or LC-MS