Learn More
Medchemexpress LLC Valproic acid-d4 | 87745-17-3 | 98.0% | 148.24 | C8H12D4O2 | 1 MG
SDP
Catalog No. 5000244291
Encompass_Preferred
Valproic acid-d4 is the deuterium-labeled analogue of valproic acid provided for analytical and research use. It functions as a stable isotope internal standard for quantitative mass spectrometry, enabling accurate measurement through a predictable mass shift and comparable chromatographic behavior. Supplied in small milligram quantities for method development, calibration, and trace-level analysis.
  • Deuterium-labeled internal standard for LC-MS and GC-MS assays.
  • Four deuterium atoms provide a distinct mass shift for accurate quantitation.
  • High purity (98.0%) for reliable analytical performance.
  • Suitable for method development, calibration, and quality control in small-mass formats (1 mg).
  • Documentation available: SDS, certificate of analysis, and NMR data.
Catalog No. 50-002-44291 Supplier Medchemexpress LLC Supplier No. HY10585S1MG
Valproic acid-d4 is the deuterium-labeled analogue of valproic acid provided for analytical and research use. It functions as a stable isotope internal standard for quantitative mass spectrometry, enabling accurate measurement through a predictable mass shift and comparable chromatographic behavior. Supplied in small milligram quantities for method development, calibration, and trace-level analysis.
  • Deuterium-labeled internal standard for LC-MS and GC-MS assays.
  • Four deuterium atoms provide a distinct mass shift for accurate quantitation.
  • High purity (98.0%) for reliable analytical performance.
  • Suitable for method development, calibration, and quality control in small-mass formats (1 mg).
  • Documentation available: SDS, certificate of analysis, and NMR data.