Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valproic acid-d4 (sodium salt) | 87745-17-3 | MFCD01311697 | 98.0% | 148.24 g/mol | C8H11D4NaO2 | 5 MG
SDP

Catalog No. 5000243296
Encompass_Preferred

Valproic acid-d4 (sodium salt) is the deuterium-labeled analogue of valproic acid, provided as a high-purity reagent for use as a stable isotope internal standard in quantitative mass spectrometry and bioanalytical assays. It is intended to improve accuracy and reproducibility in LC-MS and GC-MS method development, validation, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (98.0%) for reliable analytical performance.
  • Suitable for LC-MS and GC-MS quantitative assays.
  • Provided as the sodium salt for improved solubility in aqueous media.
  • Molecular formula C8H11D4NaO2; molecular weight 148.24 g/mol.
  • Available in small analytical pack sizes for laboratory use.

Catalog No. 50-002-43296 Supplier Medchemexpress LLC Supplier No. HY10585S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valproic acid-d4 (sodium salt) is the deuterium-labeled analogue of valproic acid, provided as a high-purity reagent for use as a stable isotope internal standard in quantitative mass spectrometry and bioanalytical assays. It is intended to improve accuracy and reproducibility in LC-MS and GC-MS method development, validation, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (98.0%) for reliable analytical performance.
  • Suitable for LC-MS and GC-MS quantitative assays.
  • Provided as the sodium salt for improved solubility in aqueous media.
  • Molecular formula C8H11D4NaO2; molecular weight 148.24 g/mol.
  • Available in small analytical pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.