Learn More
Medchemexpress LLC Valproic acid-d6 | 87745-18-4 | MFCD01074058 | 99.8% | 150.25 | C8H10D6O2 | 10 MG

Supplier: Medchemexpress LLC HY10585S110MG
Valproic acid-d6 is a deuterium-labeled analogue of valproic acid used as an internal standard and reference material for analytical, pharmacokinetic, and method development applications. Supplied as a liquid isotopically labeled standard, it provides consistent isotopic enrichment and purity for reliable mass spectrometry and quality-control workflows.
- Deuterium labeled for use as an isotope-dilution internal standard.
- High purity (99.8%) for accurate analytical results.
- Chemical formula C8H10D6O2; molecular weight 150.25.
- Available in small, precise quantities suitable for analytical use.
- Storage stability: pure form stable at -20°C (3 years) or 4°C (2 years).
- Compatible with LC-MS and GC-MS method validation and pharmacokinetic studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.