Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valproic acid-d6 | 87745-18-4 | MFCD01074058 | 99.8% | 150.25 | C8H10D6O2 | 5 MG
SDP

Catalog No. 5000243760
Encompass_Preferred

Valproic acid-d6 is the deuterium-labeled analogue of valproic acid used as an internal standard and tracer in analytical chemistry and biochemical research. It contains six deuterium atoms for clear mass differentiation and is intended for quantitative assays where isotopic labeling improves accuracy and precision.

  • deuterium-labeled internal standard (d6) for quantitative analysis
  • high isotopic purity, 99.8%
  • molecular formula C8H10D6O2; molecular weight 150.25
  • suitable for NMR, GC-MS, and LC-MS applications
  • store at -20°C for long-term; follow recommended solvent storage conditions
  • colorless to light yellow liquid appearance

Catalog No. 50-002-43760 Supplier Medchemexpress LLC Supplier No. HY10585S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valproic acid-d6 is the deuterium-labeled analogue of valproic acid used as an internal standard and tracer in analytical chemistry and biochemical research. It contains six deuterium atoms for clear mass differentiation and is intended for quantitative assays where isotopic labeling improves accuracy and precision.

  • deuterium-labeled internal standard (d6) for quantitative analysis
  • high isotopic purity, 99.8%
  • molecular formula C8H10D6O2; molecular weight 150.25
  • suitable for NMR, GC-MS, and LC-MS applications
  • store at -20°C for long-term; follow recommended solvent storage conditions
  • colorless to light yellow liquid appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.