Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valproic acid-d6 | 87745-18-4 | MFCD01074058 | 99.8% | 150.25 g/mol | C8H10D6O2 | 1 MG
SDP

Catalog No. 5000244417
Encompass_Preferred

Valproic acid-d6 is the deuterium-labeled analogue of valproic acid used as an isotopic standard and tracer in analytical and research workflows. It provides a defined mass shift for accurate quantitation by mass spectrometry and is supplied as a high-purity reagent in small-scale quantities.

  • Deuterium-labeled analogue for use as an internal standard in mass spectrometry.
  • High purity suitable for analytical applications (reported 99.8%).
  • Molecular formula C8H10D6O2; molecular weight 150.25 g/mol.
  • Supplied as a small, 1 mg package for method development and validation.
  • Store at -20°C for long-term stability; short-term storage at 4°C.

Catalog No. 50-002-44417 Supplier Medchemexpress LLC Supplier No. HY10585S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valproic acid-d6 is the deuterium-labeled analogue of valproic acid used as an isotopic standard and tracer in analytical and research workflows. It provides a defined mass shift for accurate quantitation by mass spectrometry and is supplied as a high-purity reagent in small-scale quantities.

  • Deuterium-labeled analogue for use as an internal standard in mass spectrometry.
  • High purity suitable for analytical applications (reported 99.8%).
  • Molecular formula C8H10D6O2; molecular weight 150.25 g/mol.
  • Supplied as a small, 1 mg package for method development and validation.
  • Store at -20°C for long-term stability; short-term storage at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.