Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valproic acid impurity 1 | 5343-52-2 | 97.0% | 144.21 | 250 MG
SDP

Catalog No. 5000402285
Encompass_Preferred

Valproic acid impurity 1 is an impurity of Valproic acid. It is intended for research use only as a laboratory chemical and for the manufacture of substances.

  • Purity: 97.0%
  • Store at room temperature 3 years
  • In solvent: -80°C 2 years, -20°C 1 year
  • Appearance: Liquid

Catalog No. 50-004-02285 Supplier Medchemexpress LLC Supplier No. 250MGHY33358
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valproic acid impurity 1 is an impurity of Valproic acid. It is intended for research use only as a laboratory chemical and for the manufacture of substances.

  • Purity: 97.0%
  • Store at room temperature 3 years
  • In solvent: -80°C 2 years, -20°C 1 year
  • Appearance: Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.