Learn More
Selleck Chemical LLC Valproic acid sodium salt (Sodium valproate) 200mg 1069-66-5
SDP
Catalog No. 507457
Encompass_Preferred
Valproic acid sodium salt (Sodium valproate) is a HDAC inhibitor by selectively inducing proteasomal degradation of HDAC2, used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Valproic acid induces Notch1 signaling in small cell lung cancer (SCLC) cells. Valproic acid is under investigation for treatment of HIV and various cancers. Valproic acid (VPA) induces autophagy and mitophagy by upregulation of BNIP3 and mitochondrial biogenesis by upregulating PGC-1?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-745-7 Supplier Selleck Chemical LLC Supplier No. S1168200MG
Add to Cart
Add to Cart
Valproic acid sodium salt (Sodium valproate) is a HDAC inhibitor by selectively inducing proteasomal degradation of HDAC2, used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Valproic acid induces Notch1 signaling in small cell lung cancer (SCLC) cells. Valproic acid is under investigation for treatment of HIV and various cancers. Valproic acid (VPA) induces autophagy and mitophagy by upregulation of BNIP3 and mitochondrial biogenesis by upregulating PGC-1?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.