Learn More
Medchemexpress LLC HY-10585A 5g , Valproic acid (sodium salt) Sodium Valproate CAS:1069-66-5 Purity:98%
SDP
Catalog No. 501492187
Encompass_Preferred
Medchemexpress, HY-10585A 5g Valproic acid (sodium salt) Sodium Valproate CAS:1069-66-5 Formula:C8H15NaO2 IC 50 : 400 μM (HDAC1), 0.5-2 mM (HDAC) HDAC2 [6] Purity:98% Valproic acid sodium salt is an HDAC inhibitor, with IC 50 in the range of 0.5 and 2 mM, also inhibits HDAC1 ( IC 50 , 400 μM), and induces proteasomal degradation of HDAC2 ; Valproic acid sodium salt is used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2187 Supplier Medchemexpress LLC Supplier No. HY10585A5G
Medchemexpress, HY-10585A 5g Valproic acid (sodium salt) Sodium Valproate CAS:1069-66-5 Formula:C8H15NaO2 IC 50 : 400 μM (HDAC1), 0.5-2 mM (HDAC) HDAC2 [6] Purity:98% Valproic acid sodium salt is an HDAC inhibitor, with IC 50 in the range of 0.5 and 2 mM, also inhibits HDAC1 ( IC 50 , 400 μM), and induces proteasomal degradation of HDAC2 ; Valproic acid sodium salt is used in the treatment of epilepsy, bipolar disorder and prevention of migraine headaches. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.