Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valproic Acid, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607685 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-685 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-685 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V201125GM

missing translation for 'validMainframeMsg'

V2011, 99-66-1, C8H16O2

Spectrum™ Chemical Valproic Acid, USP is used to treat certain types of seizures. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 99-66-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00002672
InChI Key NIJJYAXOARWZEE-UHFFFAOYSA-N
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.