Learn More
Medchemexpress LLC HY-B2117 10mg Medchemexpress, Valpromide CAS:2430-27-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB211710MG
Encompass_Preferred
Medchemexpress, HY-B2117 10mg Valpromide CAS:2430-27-5 Valpromide is an amide derivative of valproic acid and inhibits human epoxide hydrolase . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3067