Learn More
Cayman Chemical Valrocemide, Purity: ≥95%, CAS: 92262-58-3, 100 mg
SDP
Catalog No. 502990483
Encompass_Preferred
An anticonvulsant; protects against MES-, PTZ-, picrotoxin-, or bicuculline-induced seizures in mice (ED50s = 151, 132, 275, and 248 mg/kg, respectively); prevents sound-induced seizures in Frings audiogenic-sensitive mice (ED50 = 52 mg/kg) and MES-induced seizures in rats (ED50 = 73 mg/kg); prevents seizures in corneally kindled rats (ED50 = 161 mg/kg)
Catalog No. 50-299-0483 Supplier Cayman Chemical Supplier No. 30909100 MG
An anticonvulsant; protects against MES-, PTZ-, picrotoxin-, or bicuculline-induced seizures in mice (ED50s = 151, 132, 275, and 248 mg/kg, respectively); prevents sound-induced seizures in Frings audiogenic-sensitive mice (ED50 = 52 mg/kg) and MES-induced seizures in rats (ED50 = 73 mg/kg); prevents seizures in corneally kindled rats (ED50 = 161 mg/kg)