Learn More
Cayman Chemical ValrocemIde 50mg
SDP
Catalog No. 502990295
Encompass_Preferred
An anticonvulsant; protects against MES-, PTZ-, picrotoxin-, or bicuculline-induced seizures in mice (ED50s = 151, 132, 275, and 248 mg/kg, respectively); prevents sound-induced seizures in Frings audiogenic-sensitive mice (ED50 = 52 mg/kg) and MES-induced seizures in rats (ED50 = 73 mg/kg); prevents seizures in corneally kindled rats (ED50 = 161 mg/kg)
Catalog No. 50-299-0295 Supplier Cayman Chemical Supplier No. 3090950 MG
An anticonvulsant; protects against MES-, PTZ-, picrotoxin-, or bicuculline-induced seizures in mice (ED50s = 151, 132, 275, and 248 mg/kg, respectively); prevents sound-induced seizures in Frings audiogenic-sensitive mice (ED50 = 52 mg/kg) and MES-induced seizures in rats (ED50 = 73 mg/kg); prevents seizures in corneally kindled rats (ED50 = 161 mg/kg)