Learn More
Medchemexpress LLC Pentanoic acid, 2-[(2S,4S)-1,2,3,4,6,11-hexahydro-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-4-[ | 56124-62-0 | MFCD01939322 | 99.6% | C34H36F3NO13 | 10MG
SDP
Catalog No. 501872957
Encompass_Preferred
Valrubicin is an anthracycline chemotherapy agent provided for research use. It inhibits TPA- and PDBu-induced protein kinase C (PKC) activation with reported IC50 values of 0.85 μM and 1.25 μM, and demonstrates antitumor and anti-inflammatory activity in preclinical studies.
  • Inhibits TPA- and PDBu-induced protein kinase C activation (IC50 0.85 μM and 1.25 μM).
  • Exhibits antitumor and anti-inflammatory activity in preclinical models.
  • Provided as a research-grade small molecule for biochemical and cellular assays.
  • High purity suitable for analytical applications (purity 99.6% by LCMS).
  • Available in small pack sizes suitable for assay development and reference standards.
Catalog No. 50-187-2957 Supplier Medchemexpress LLC Supplier No. HY1377210MG
Valrubicin is an anthracycline chemotherapy agent provided for research use. It inhibits TPA- and PDBu-induced protein kinase C (PKC) activation with reported IC50 values of 0.85 μM and 1.25 μM, and demonstrates antitumor and anti-inflammatory activity in preclinical studies.
  • Inhibits TPA- and PDBu-induced protein kinase C activation (IC50 0.85 μM and 1.25 μM).
  • Exhibits antitumor and anti-inflammatory activity in preclinical models.
  • Provided as a research-grade small molecule for biochemical and cellular assays.
  • High purity suitable for analytical applications (purity 99.6% by LCMS).
  • Available in small pack sizes suitable for assay development and reference standards.