Learn More
Med Vet International Amber Plastic Graduated Ovals, 16oz, J1207F, 50/Case
Catalog No. 5000495700
Encompass_Preferred
Amber plastic graduated ovals with twist off child resistant caps. Light-weight and transparent. Graduated in mls 16oz 50/case
Catalog No. 50-004-95700 Supplier Med Vet International Supplier No. JOR1207F
Amber plastic graduated ovals with twist off child resistant caps. Light-weight and transparent. Graduated in mls 16oz 50/case