Learn More
Medchemexpress LLC Valsartan-d9 | 1089736-73-1 | 99.3% | 444.57 | 1 MG
SDP
Catalog No. 5000387547
Encompass_Preferred
Valsartan-d9 is deuterium labeled valsartan, an angiotensin II receptor antagonist with potential applications in high blood pressure and heart failure research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87547 Supplier Medchemexpress LLC Supplier No. HY18204S1MG
Valsartan-d9 is deuterium labeled valsartan, an angiotensin II receptor antagonist with potential applications in high blood pressure and heart failure research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.