Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valsartan ethyl ester | 1111177-30-0 | MFCD29045635 | 99.7% | C26H33N5O3 | 10MG
SDP

Catalog No. 5000259805
Encompass_Preferred

Valsartan ethyl ester is an impurity and analytical reference related to valsartan, an angiotensin II receptor antagonist used in hypertension and heart failure research. Supplied for research and analytical testing, it is characterized by high reported purity and specified storage recommendations.

  • High purity (99.7%).
  • Molecular formula C26H33N5O3.
  • Molecular weight 463.57 g/mol.
  • Suitable as an analytical reference or impurity standard.
  • Available in small pack sizes (5 mg, 10 mg, 50 mg, 100 mg).
  • Store protected from light; in solvent, -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-59805 Supplier Medchemexpress LLC Supplier No. HY13536310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valsartan ethyl ester is an impurity and analytical reference related to valsartan, an angiotensin II receptor antagonist used in hypertension and heart failure research. Supplied for research and analytical testing, it is characterized by high reported purity and specified storage recommendations.

  • High purity (99.7%).
  • Molecular formula C26H33N5O3.
  • Molecular weight 463.57 g/mol.
  • Suitable as an analytical reference or impurity standard.
  • Available in small pack sizes (5 mg, 10 mg, 50 mg, 100 mg).
  • Store protected from light; in solvent, -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.