Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valsartan Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013017 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3017 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3017 Supplier MilliporeSigma Supelco Supplier No. PHR13151G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Valsartan is an orally active specific angiotensin II receptor blocker used for the treatment of hypertension.

TRUSTED_SUSTAINABILITY

Specifications

CAS 137862-53-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C24H29N5O3
MDL Number MFCD00865840
Synonym Valsartan; N-(1-Oxopentyl)-N-[[2′-(2 H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine
Type Certified Reference Material
Quantity 1 g
Formula Weight 435.52
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.