Learn More
Medchemexpress LLC Valspodar | 121584-18-7 | 99.3% | 1214.62 | 1 MG
SDP
Catalog No. 5000384958
Encompass_Preferred
Valspodar (PSC 833) is a selective P-glycoprotein inhibitor that has been used as an experimental cancer treatment and chemosensitizer.
  • Solid appearance, white to yellow color
  • Ships at room temperature in continental US
  • Store powder at -20°C for 3 years, or in solvent at -80°C for 6 months / -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL (82.33 mM)
  • Exhibits no cytotoxicity effects up to 0.75 μg/mL
  • Increases cell kill efficacy of MDR cell type when administered with DOX-L
  • Displays slow clearance and large volume of distribution in vivo
  • Shows low hepatic extraction and wide distribution similar to CsA in vivo
Catalog No. 50-003-84958 Supplier Medchemexpress LLC Supplier No. HY173841MG
Valspodar (PSC 833) is a selective P-glycoprotein inhibitor that has been used as an experimental cancer treatment and chemosensitizer.
  • Solid appearance, white to yellow color
  • Ships at room temperature in continental US
  • Store powder at -20°C for 3 years, or in solvent at -80°C for 6 months / -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL (82.33 mM)
  • Exhibits no cytotoxicity effects up to 0.75 μg/mL
  • Increases cell kill efficacy of MDR cell type when administered with DOX-L
  • Displays slow clearance and large volume of distribution in vivo
  • Shows low hepatic extraction and wide distribution similar to CsA in vivo