Learn More
Abcam Valspodar (PSC-833), P-glycoprotein inhibitor, 1MG
Catalog No. 502705053
Encompass_Preferred
MW 1214.6 Da, Purity >94%. Selective P-glycoprotein inhibitor. Cyclosporin (ab143599) analog. Shows chemosensitizing effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5053 Supplier Abcam Supplier No. AB1458701MG
MW 1214.6 Da, Purity >94%. Selective P-glycoprotein inhibitor. Cyclosporin (ab143599) analog. Shows chemosensitizing effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement