Learn More
MSC Value Collection 12 in Sealer Repair Kit, 1 ea
Catalog No. 5000795221
Encompass
This repair kit provides essential components for maintaining tabletop thermal sealers. It is designed to restore functionality by replacing worn parts.
  • Designed for tabletop thermal sealers
  • Maximum seal size of 12 in
  • Includes one lower PTFE tape
  • Includes two upper PTFE tapes
  • Includes two wire elements
Catalog No. 50-007-95221 Supplier MSC Supplier No. 76218965
This repair kit provides essential components for maintaining tabletop thermal sealers. It is designed to restore functionality by replacing worn parts.
  • Designed for tabletop thermal sealers
  • Maximum seal size of 12 in
  • Includes one lower PTFE tape
  • Includes two upper PTFE tapes
  • Includes two wire elements