Learn More
MSC Value Collection Aircraft Cable, 1/16 in, 1 x 19 Single Strand, Stainless Steel
Catalog No. 5000527692
Encompass
This aircraft cable is a durable and reliable solution for various industrial and commercial applications. Constructed from stainless steel with a 1/16 inch diameter and 1 x 19 single strand configuration, it offers excellent strength and corrosion resistance. It is designed to meet demanding performance requirements.
  • 1/16 inch diameter for versatile applications.
  • 1 x 19 single strand construction provides high strength.
  • Stainless steel finish ensures corrosion resistance and durability.
  • 500 lb breaking strength for heavy-duty use.
  • Uncoated for general purpose applications.
  • Supplied in a 500 ft roll.
Catalog No. 50-005-27692 Supplier MSC Supplier No. 45701174
This aircraft cable is a durable and reliable solution for various industrial and commercial applications. Constructed from stainless steel with a 1/16 inch diameter and 1 x 19 single strand configuration, it offers excellent strength and corrosion resistance. It is designed to meet demanding performance requirements.
  • 1/16 inch diameter for versatile applications.
  • 1 x 19 single strand construction provides high strength.
  • Stainless steel finish ensures corrosion resistance and durability.
  • 500 lb breaking strength for heavy-duty use.
  • Uncoated for general purpose applications.
  • Supplied in a 500 ft roll.