Learn More
MSC Value Collection Aircraft Cable, 1/4 in Diameter, 7x7 Strand Core, Galvanized
Catalog No. 5000528081
Encompass
This aircraft cable is designed with a 1/4 inch diameter and a 7 x 7 strand core. It offers a breaking strength of 6,100 Lb and is supplied in a 500-foot roll. The cable features a galvanized finish and is uncoated, providing durability for a range of uses.
  • Breaking strength: 6,100 Lb
  • Bare diameter: 1/4 in
  • Length: 500 ft
  • Finish: Galvanized
  • Strand type: 7 x 7 strand core
  • Head coating: Uncoated
Catalog No. 50-005-28081 Supplier MSC Supplier No. 45701851
This aircraft cable is designed with a 1/4 inch diameter and a 7 x 7 strand core. It offers a breaking strength of 6,100 Lb and is supplied in a 500-foot roll. The cable features a galvanized finish and is uncoated, providing durability for a range of uses.
  • Breaking strength: 6,100 Lb
  • Bare diameter: 1/4 in
  • Length: 500 ft
  • Finish: Galvanized
  • Strand type: 7 x 7 strand core
  • Head coating: Uncoated