Learn More
MSC Value Collection Aircraft Cable, 3/32 in Coated, Galvanized, 480 lb
Catalog No. 5000528219
Encompass
This aircraft cable is constructed from galvanized material and features a protective vinyl coating. Designed with a 7 x 7 strand core, it offers a robust breaking strength of 480 lb and a coated diameter of 3/32 in, making it suitable for various demanding applications.
  • Galvanized construction
  • Vinyl coating
  • 7 x 7 strand core
  • 480 lb breaking strength
  • 3/32 in coated diameter
  • 1/16 in bare diameter
  • Priced as 1 ft increments
Catalog No. 50-005-28219 Supplier MSC Supplier No. 45700812
This aircraft cable is constructed from galvanized material and features a protective vinyl coating. Designed with a 7 x 7 strand core, it offers a robust breaking strength of 480 lb and a coated diameter of 3/32 in, making it suitable for various demanding applications.
  • Galvanized construction
  • Vinyl coating
  • 7 x 7 strand core
  • 480 lb breaking strength
  • 3/32 in coated diameter
  • 1/16 in bare diameter
  • Priced as 1 ft increments