Learn More
MSC Value Collection Aircraft Cable, 5/16 in, 8,000 lb, Galvanized Steel
Catalog No. 5000536837
Encompass
This aircraft cable features a 5/16 inch diameter and a total coil length of 1,000 feet. It is designed with a 1 x 7 single strand construction, offers an 8,000 lb breaking strength, and has a galvanized finish for durability.
  • 5/16 inch diameter
  • 1,000 foot total coil length
  • 8,000 lb breaking strength
  • 1 x 7 single strand construction
  • Galvanized finish
  • Priced in 1 foot increments
Catalog No. 50-005-36837 Supplier MSC Supplier No. 89489082
This aircraft cable features a 5/16 inch diameter and a total coil length of 1,000 feet. It is designed with a 1 x 7 single strand construction, offers an 8,000 lb breaking strength, and has a galvanized finish for durability.
  • 5/16 inch diameter
  • 1,000 foot total coil length
  • 8,000 lb breaking strength
  • 1 x 7 single strand construction
  • Galvanized finish
  • Priced in 1 foot increments