Learn More
MSC Value Collection All Purpose Snap, 80 lb Capacity, Steel, Zinc Finish
Catalog No. 5000510228
Encompass
This all purpose snap is constructed from steel and features a zinc finish, providing durability and corrosion resistance. It offers a working load capacity of 80 lb, making it suitable for various general-purpose applications.
  • All purpose snap
  • 80 lb working load capacity
  • 320 lb breaking load capacity
  • Steel construction
  • Zinc finish
  • 3/16 in size
Catalog No. 50-005-10228 Supplier MSC Supplier No. 67791764
This all purpose snap is constructed from steel and features a zinc finish, providing durability and corrosion resistance. It offers a working load capacity of 80 lb, making it suitable for various general-purpose applications.
  • All purpose snap
  • 80 lb working load capacity
  • 320 lb breaking load capacity
  • Steel construction
  • Zinc finish
  • 3/16 in size