Learn More
MSC Value Collection Angle Block Set, 1-30° Angle, 3 in Long, 45-50 HRC
Catalog No. 5000529359
Encompass
This angle block set provides precise angle measurement and setup capabilities for various applications. It features a robust construction with a specific hardness to ensure durability and accuracy. The comprehensive 8-piece set includes both angle plates and V-blocks, covering a wide range of angles.
  • Hardness of 45-50 HRC
  • Includes three V-blocks at 10, 15, and 30 degrees
  • Includes five angle plates covering 1 to 5 degrees
  • Total of eight pieces for versatile use
  • Minimum angle included is 1 degree
  • Maximum angle included is 30 degrees
Catalog No. 50-005-29359 Supplier MSC Supplier No. 06366108
This angle block set provides precise angle measurement and setup capabilities for various applications. It features a robust construction with a specific hardness to ensure durability and accuracy. The comprehensive 8-piece set includes both angle plates and V-blocks, covering a wide range of angles.
  • Hardness of 45-50 HRC
  • Includes three V-blocks at 10, 15, and 30 degrees
  • Includes five angle plates covering 1 to 5 degrees
  • Total of eight pieces for versatile use
  • Minimum angle included is 1 degree
  • Maximum angle included is 30 degrees