Learn More
MSC Value Collection Ball Transfer, Chrome Steel, Round Base, 7/8 in
Catalog No. 5000529825
Encompass
This ball transfer unit provides smooth, multi-directional movement for various applications. It features chrome steel balls, a round base, and a durable steel housing, designed for efficient material handling.
  • Round base shape
  • Mount height: 7/8 in
  • Ball material: Chrome steel
  • Thread size: 5/16-24
  • Thread length: 1 in
  • Overall diameter: 7/8 in
  • Housing material: Steel
Catalog No. 50-005-29825 Supplier MSC Supplier No. 00290825
This ball transfer unit provides smooth, multi-directional movement for various applications. It features chrome steel balls, a round base, and a durable steel housing, designed for efficient material handling.
  • Round base shape
  • Mount height: 7/8 in
  • Ball material: Chrome steel
  • Thread size: 5/16-24
  • Thread length: 1 in
  • Overall diameter: 7/8 in
  • Housing material: Steel