Learn More
MSC Value Collection Ball Transfer, Chrome Steel, Square Base, 2 in Diameter
Catalog No. 5000530002
Encompass
This ball transfer unit features a chrome steel ball and a square base, designed for efficient material handling. With a 2 in overall diameter and a 1/2 in mount height, it is suitable for various applications requiring smooth, multi-directional movement.
  • Chrome steel ball for durability
  • Square base for stable mounting
  • 2 in overall diameter facilitates easy movement
  • 1/2 in mount height for versatile integration
  • Steel housing material provides robust construction
Catalog No. 50-005-30002 Supplier MSC Supplier No. 00290940
This ball transfer unit features a chrome steel ball and a square base, designed for efficient material handling. With a 2 in overall diameter and a 1/2 in mount height, it is suitable for various applications requiring smooth, multi-directional movement.
  • Chrome steel ball for durability
  • Square base for stable mounting
  • 2 in overall diameter facilitates easy movement
  • 1/2 in mount height for versatile integration
  • Steel housing material provides robust construction