Learn More
MSC Value Collection Bevel Protractor, 6 in Blade, 360 Degree Measurement, 5 Minute Accuracy
Catalog No. 5000517003
Encompass
This bevel protractor is equipped with a 6 inch long blade, providing a maximum measurement range of 360 degrees. It delivers high precision with an accuracy of 5 minutes and a dial graduation of 5 minutes.
  • Features a 6 inch long blade
  • Offers a maximum measurement range of 360 degrees
  • Provides an accuracy of 5 minutes
  • Has a dial graduation of 5 minutes
  • Includes a dial reading of 0-90-0
Catalog No. 50-005-17003 Supplier MSC Supplier No. 06476956
This bevel protractor is equipped with a 6 inch long blade, providing a maximum measurement range of 360 degrees. It delivers high precision with an accuracy of 5 minutes and a dial graduation of 5 minutes.
  • Features a 6 inch long blade
  • Offers a maximum measurement range of 360 degrees
  • Provides an accuracy of 5 minutes
  • Has a dial graduation of 5 minutes
  • Includes a dial reading of 0-90-0