Learn More
MSC Value Collection Boltless Cross Beam, Steel, 72 in Wide, 16 Gauge
Catalog No. 5000528655
Encompass
The Value Collection Boltless Cross Beam is engineered for compatibility with boltless storage racks. Crafted from robust steel and finished with a gray powder coating, this cross beam offers durable and stable support for a wide range of storage applications.
  • Designed for use with boltless storage racks.
  • Constructed from steel.
  • Features a durable gray powder-coated finish.
  • Measures 72 in wide.
  • Provides a 26 in height.
  • Made with 16 gauge material.
Catalog No. 50-005-28655 Supplier MSC Supplier No. 41525320
The Value Collection Boltless Cross Beam is engineered for compatibility with boltless storage racks. Crafted from robust steel and finished with a gray powder coating, this cross beam offers durable and stable support for a wide range of storage applications.
  • Designed for use with boltless storage racks.
  • Constructed from steel.
  • Features a durable gray powder-coated finish.
  • Measures 72 in wide.
  • Provides a 26 in height.
  • Made with 16 gauge material.