Learn More
MSC Value Collection Bore Gage Anvil Set, 8 Piece, 0.7 to 1.5 Inch Measurement Range
Catalog No. 5000516779
Encompass
This bore gage anvil set is designed for use with dial bore gages. It includes 8 pieces and provides a measurement range of 0.7 to 1.5 inches. This set offers a quality solution for precise measurements.
  • Designed for use with dial bore gages
  • Includes 8 pieces
  • Offers a measurement range of 0.7 to 1.5 inches
Catalog No. 50-005-16779 Supplier MSC Supplier No. 06416127
This bore gage anvil set is designed for use with dial bore gages. It includes 8 pieces and provides a measurement range of 0.7 to 1.5 inches. This set offers a quality solution for precise measurements.
  • Designed for use with dial bore gages
  • Includes 8 pieces
  • Offers a measurement range of 0.7 to 1.5 inches