Learn More
MSC Value Collection Braided Seine Twine, #15, Nylon, White, 132 lb Breaking Strength
Catalog No. 5000527696
Encompass
This braided seine twine is made from durable nylon and comes in a white color. It features a #15 twine size and offers a robust breaking strength of 132 lb. Ideal for various applications requiring strong and reliable cordage.
  • Made from durable nylon material
  • White color for visibility or specific applications
  • #15 twine size for versatile use
  • Offers 132 lb breaking strength for reliable performance
  • Provides 1000 ft of twine per pound
  • Supplied as a 500 foot piece for convenience
Catalog No. 50-005-27696 Supplier MSC Supplier No. 45890522
This braided seine twine is made from durable nylon and comes in a white color. It features a #15 twine size and offers a robust breaking strength of 132 lb. Ideal for various applications requiring strong and reliable cordage.
  • Made from durable nylon material
  • White color for visibility or specific applications
  • #15 twine size for versatile use
  • Offers 132 lb breaking strength for reliable performance
  • Provides 1000 ft of twine per pound
  • Supplied as a 500 foot piece for convenience