Learn More
MSC Value Collection Braided Seine Twine, Nylon, #15 Twine Dia, Pink
Catalog No. 5000534787
Encompass
This braided seine twine is made from durable nylon and features a #15 twine diameter, making it suitable for various applications requiring strength and reliability. It comes in a distinct pink color.
  • Made from durable nylon material
  • #15 Twine diameter
  • Pink color
  • Breaking strength of 132 lb
  • Provides 1,000 ft of twine per pound
Catalog No. 50-005-34787 Supplier MSC Supplier No. 45890563
This braided seine twine is made from durable nylon and features a #15 twine diameter, making it suitable for various applications requiring strength and reliability. It comes in a distinct pink color.
  • Made from durable nylon material
  • #15 Twine diameter
  • Pink color
  • Breaking strength of 132 lb
  • Provides 1,000 ft of twine per pound