Learn More
MSC Value Collection Caster, Swivel, Phenolic, 4 in Wheel, 800 lb
Catalog No. 5000510503
Encompass
This swivel top plate caster features a phenolic wheel with a 4 inch diameter and 2 inch width, offering an 800 lb load capacity. It is designed for general purpose applications, providing reliable mobility.
  • Roller bearing for smooth operation.
  • Stamp formed construction.
  • Powder coated finish.
  • Top plate mount.
  • General purpose application.
  • Phenolic wheel material.
  • Overall height of 5-5/8 inches.
  • Plate size of 4 x 4-1/2 inches.
Catalog No. 50-005-10503 Supplier MSC Supplier No. 02276889
This swivel top plate caster features a phenolic wheel with a 4 inch diameter and 2 inch width, offering an 800 lb load capacity. It is designed for general purpose applications, providing reliable mobility.
  • Roller bearing for smooth operation.
  • Stamp formed construction.
  • Powder coated finish.
  • Top plate mount.
  • General purpose application.
  • Phenolic wheel material.
  • Overall height of 5-5/8 inches.
  • Plate size of 4 x 4-1/2 inches.