Learn More
MSC Value Collection Clevis Hook, 1/4 in Chain, Grade 70
Catalog No. 5000528174
Encompass
This clevis hook is designed for secure connections in various applications, featuring a 1/4 inch chain diameter and a 70 chain grade. It is constructed from durable alloy steel, offering robust performance and reliable load handling.
  • 1/4 inch chain diameter
  • 70 chain grade
  • 3,150 Lb capacity
  • 3/4 inch hook throat
  • 2-39/64 inch overall length
  • Made from alloy steel
Catalog No. 50-005-28174 Supplier MSC Supplier No. 53590055
This clevis hook is designed for secure connections in various applications, featuring a 1/4 inch chain diameter and a 70 chain grade. It is constructed from durable alloy steel, offering robust performance and reliable load handling.
  • 1/4 inch chain diameter
  • 70 chain grade
  • 3,150 Lb capacity
  • 3/4 inch hook throat
  • 2-39/64 inch overall length
  • Made from alloy steel