Learn More
MSC Value Collection Clevis Hook, 3/8 in Chain Dia, 100 Chain Grade, 8,800 Lb Capacity
Catalog No. 5000514786
Encompass
This Clevis Hook from Value Collection is designed for heavy-duty lifting applications, featuring a durable alloy steel construction and a high capacity.
  • Designed for heavy-duty lifting applications.
  • Constructed from durable alloy steel.
  • Features a 100 chain grade.
  • Provides an 8,800 Lb (3,991.6 kg) capacity.
  • Has a clevis pin diameter of 9/16 in (14.29 mm).
  • Inside diameter is 1-9/32 in (32.54 mm).
  • Hook throat measures 1-5/8 in (41.28 mm).
  • Overall length is 4-13/16 in (122.24 mm).
Catalog No. 50-005-14786 Supplier MSC Supplier No. 53595344
This Clevis Hook from Value Collection is designed for heavy-duty lifting applications, featuring a durable alloy steel construction and a high capacity.
  • Designed for heavy-duty lifting applications.
  • Constructed from durable alloy steel.
  • Features a 100 chain grade.
  • Provides an 8,800 Lb (3,991.6 kg) capacity.
  • Has a clevis pin diameter of 9/16 in (14.29 mm).
  • Inside diameter is 1-9/32 in (32.54 mm).
  • Hook throat measures 1-5/8 in (41.28 mm).
  • Overall length is 4-13/16 in (122.24 mm).