Learn More
MSC Value Collection Clevis Hook, 5/16 inch Chain Diameter, Grade 43, 3,900 lb Capacity
Catalog No. 5000528610
Encompass
This clevis hook is designed for secure lifting applications, featuring a 5/16 inch chain diameter and a Grade 43 rating. It offers a robust 3,900 lb capacity, along with a 1/2 inch hook throat and an overall length of 2-7/8 inches.
  • 5/16 inch chain diameter
  • Grade 43 chain
  • 3,900 lb capacity
  • 1/2 inch hook throat
  • 2-7/8 inch overall length
  • Made from alloy steel
Catalog No. 50-005-28610 Supplier MSC Supplier No. 53589966
This clevis hook is designed for secure lifting applications, featuring a 5/16 inch chain diameter and a Grade 43 rating. It offers a robust 3,900 lb capacity, along with a 1/2 inch hook throat and an overall length of 2-7/8 inches.
  • 5/16 inch chain diameter
  • Grade 43 chain
  • 3,900 lb capacity
  • 1/2 inch hook throat
  • 2-7/8 inch overall length
  • Made from alloy steel