Learn More
MSC Value Collection Coiled Spring Pin, 0.125 in Dia, 3/8 in Long, 1070-1090 Alloy Steel
Catalog No. 5000508789
Encompass
This coiled spring pin is designed for various industrial applications. Made from durable 1070-1090 alloy steel, it features a plain finish and precise dimensions, ensuring reliable performance in assemblies.
  • Coiled pin type
  • 3/8 inch overall length
  • 1/8 inch diameter
  • Constructed from alloy steel
  • Features a 1070-1090 material grade
  • Plain finish
  • Inch system of measurement
  • Manufactured in the United States
Catalog No. 50-005-08789 Supplier MSC Supplier No. 67501882
This coiled spring pin is designed for various industrial applications. Made from durable 1070-1090 alloy steel, it features a plain finish and precise dimensions, ensuring reliable performance in assemblies.
  • Coiled pin type
  • 3/8 inch overall length
  • 1/8 inch diameter
  • Constructed from alloy steel
  • Features a 1070-1090 material grade
  • Plain finish
  • Inch system of measurement
  • Manufactured in the United States