Learn More
MSC Value Collection Compression Spring, 15/32 in OD, 0.0550 in Wire, 1.4690 in Free Length
Catalog No. 5000535109
Encompass
This compression spring is designed to provide reliable performance for a variety of applications. It is part of a collection known for offering quality products at a great value.
  • Outside diameter of 15/32 in
  • Wire size of 0.0550 in
  • Free length of 1.4690 in
  • Constructed from spring steel
Catalog No. 50-005-35109 Supplier MSC Supplier No. 53614442
This compression spring is designed to provide reliable performance for a variety of applications. It is part of a collection known for offering quality products at a great value.
  • Outside diameter of 15/32 in
  • Wire size of 0.0550 in
  • Free length of 1.4690 in
  • Constructed from spring steel