Learn More
MSC Value Collection Compression Spring, Spring Steel, 0.120 Wire, 1/32 OD
Catalog No. 5000528176
Encompass
This compression spring is constructed from durable spring steel, designed to resist compressive forces. It features a 0.120 inch wire size and a 1/32 inch outside diameter, making it suitable for a variety of mechanical applications where consistent force and resilience are required.
  • Constructed from spring steel.
  • Features a 0.120 inch wire size.
  • Has a 1/32 inch outside diameter.
  • Offers a free length of 2-3/16 in.
  • Provides a spring rate of 110.0000.
Catalog No. 50-005-28176 Supplier MSC Supplier No. 53614228
This compression spring is constructed from durable spring steel, designed to resist compressive forces. It features a 0.120 inch wire size and a 1/32 inch outside diameter, making it suitable for a variety of mechanical applications where consistent force and resilience are required.
  • Constructed from spring steel.
  • Features a 0.120 inch wire size.
  • Has a 1/32 inch outside diameter.
  • Offers a free length of 2-3/16 in.
  • Provides a spring rate of 110.0000.