Learn More
MSC Value Collection Dial Bore Gage, 0.24 to 0.4 in, 9 Anvils
Catalog No. 5000516809
Encompass
This dial bore gage set is designed for precise internal diameter measurements within the range of 0.24 to 0.4 inches. It provides a quality solution for various measurement needs, ensuring accuracy and reliability in bore measurements.
  • Includes 9 anvils for versatile measurement.
  • Indicator included for immediate use.
  • Comes with a plastic storage case for protection and portability.
  • Accessories included for comprehensive functionality.
Catalog No. 50-005-16809 Supplier MSC Supplier No. 06451702
This dial bore gage set is designed for precise internal diameter measurements within the range of 0.24 to 0.4 inches. It provides a quality solution for various measurement needs, ensuring accuracy and reliability in bore measurements.
  • Includes 9 anvils for versatile measurement.
  • Indicator included for immediate use.
  • Comes with a plastic storage case for protection and portability.
  • Accessories included for comprehensive functionality.