Learn More
MSC Value Collection Dial Drop Indicator, 1.0000 in Max Measurement, 0-100 Dial Reading
Catalog No. 5000535945
Encompass
The Value Collection Dial Drop Indicator is designed for precise measurement applications. It features a 1.0000 in maximum measurement range and a 0-100 dial reading with a 0.001 in dial graduation, making it suitable for detailed inspections.
  • Maximum measurement 1.0000 in
  • Dial graduation 0.001 in
  • Dial reading 0-100
  • Lug back type
  • White dial color
  • Includes revolution counter
  • English unit of measure
Catalog No. 50-005-35945 Supplier MSC Supplier No. 76450071
The Value Collection Dial Drop Indicator is designed for precise measurement applications. It features a 1.0000 in maximum measurement range and a 0-100 dial reading with a 0.001 in dial graduation, making it suitable for detailed inspections.
  • Maximum measurement 1.0000 in
  • Dial graduation 0.001 in
  • Dial reading 0-100
  • Lug back type
  • White dial color
  • Includes revolution counter
  • English unit of measure