Learn More
MSC Value Collection Dial Height Gage, 300mm, Stainless Steel, 0.02mm Graduation
Catalog No. 5000532627
Encompass
This dial height gage is designed for precise measurement applications. It features a durable stainless steel construction and a clear dial display, making it suitable for a variety of laboratory and industrial settings.
  • Graduation of 0.02 mm for fine measurements.
  • Durable stainless steel material.
  • Dial display for easy reading.
  • Used for reading fine graduations of measurement.
  • Manually adjustable for precise control.
  • Minimum measurement of 0 mm.
Catalog No. 50-005-32627 Supplier MSC Supplier No. 09834805
This dial height gage is designed for precise measurement applications. It features a durable stainless steel construction and a clear dial display, making it suitable for a variety of laboratory and industrial settings.
  • Graduation of 0.02 mm for fine measurements.
  • Durable stainless steel material.
  • Dial display for easy reading.
  • Used for reading fine graduations of measurement.
  • Manually adjustable for precise control.
  • Minimum measurement of 0 mm.