Learn More
MSC Value Collection Dial Height Gage, 6 inch, Stainless Steel, 0.001 Graduation
Catalog No. 5000513149
Encompass
This dial height gage is constructed from stainless steel and features a 6 inch measuring capacity with 0.001 inch graduation. Designed for precise measurements, it utilizes a dial display and is adjusted manually, making it suitable for reading fine graduations.
  • Display type: dial.
  • Material: stainless steel.
  • Graduation: 0.001 "
  • Used for reading fine graduations of measurement.
  • Adjusted manually.
Catalog No. 50-005-13149 Supplier MSC Supplier No. 09834771
This dial height gage is constructed from stainless steel and features a 6 inch measuring capacity with 0.001 inch graduation. Designed for precise measurements, it utilizes a dial display and is adjusted manually, making it suitable for reading fine graduations.
  • Display type: dial.
  • Material: stainless steel.
  • Graduation: 0.001 "
  • Used for reading fine graduations of measurement.
  • Adjusted manually.