Learn More
MSC Value Collection Digital Coating Thickness Gage, 0 to 40 mil, For Ferrous Bases
Catalog No. 5000530812
Encompass
This digital coating thickness gage is designed for measuring coating thickness from 0 to 40 mil on ferrous bases. It features a digital display for clear readings.
  • Measures coating thickness from 0 to 40 mil
  • Designed for use with ferrous bases
  • Features a digital display
  • Includes 2 AAA alkaline batteries
  • Comes with a long form certificate of calibration traceable to PTB
  • Includes a hard shell storage case and wrist strap
  • Includes precision plastic shims and instructions
  • Complies with multiple industry standards including ASTM B244/B499/D7091/E376, SSPC-PA2, and ISO 19840
Catalog No. 50-005-30812 Supplier MSC Supplier No. 66045477
This digital coating thickness gage is designed for measuring coating thickness from 0 to 40 mil on ferrous bases. It features a digital display for clear readings.
  • Measures coating thickness from 0 to 40 mil
  • Designed for use with ferrous bases
  • Features a digital display
  • Includes 2 AAA alkaline batteries
  • Comes with a long form certificate of calibration traceable to PTB
  • Includes a hard shell storage case and wrist strap
  • Includes precision plastic shims and instructions
  • Complies with multiple industry standards including ASTM B244/B499/D7091/E376, SSPC-PA2, and ISO 19840