Learn More
MSC Value Collection Disposable Gloves, Large, 3 mil, Nitrile, Powder-Free, 1000/Case
Catalog No. 5000787482
Encompass
These general purpose disposable gloves are made from nitrile and designed for a wide range of applications requiring hand protection. They feature a 3 mil nominal palm thickness and are ambidextrous for ease of use.
  • Nitrile construction for chemical resistance and durability
  • Three mil nominal palm thickness for tactile sensitivity and protection
  • Textured grip surface for enhanced handling
  • Powder-free to minimize contamination
  • Ambidextrous design for convenient use
  • Rolled cuff for easy donning and doffing
  • FDA approved for general purpose applications
  • Length of 9.5 in. provides extended wrist coverage
Catalog No. 50-007-87482 Supplier MSC Supplier No. 11426814
These general purpose disposable gloves are made from nitrile and designed for a wide range of applications requiring hand protection. They feature a 3 mil nominal palm thickness and are ambidextrous for ease of use.
  • Nitrile construction for chemical resistance and durability
  • Three mil nominal palm thickness for tactile sensitivity and protection
  • Textured grip surface for enhanced handling
  • Powder-free to minimize contamination
  • Ambidextrous design for convenient use
  • Rolled cuff for easy donning and doffing
  • FDA approved for general purpose applications
  • Length of 9.5 in. provides extended wrist coverage